Assay Details
| Gene Symbol: C4H6orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553589.2 | XP_005553646.2 | 1 - 2 | 186 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C4H6orf15| Gene Symbol: C4H6orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553589.2 | XP_005553646.2 | 1 - 2 | 186 | 91 | |
Set Membership: |
|