Assay Details
| Gene Symbol: GPX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287642.1 | NP_001274571.1 | 1 - 2 | 172 | 74 | |
| XM_015448838.1 | XP_015304324.1 | 1 - 2 | 440 | 74 | ||
| GenBank mRNA | X62403.1 | - | 172 | 74 | ||
More Information
Set Membership: |
|

GPX5
Genomic Map