Assay Details
| Gene Symbol: NR2F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557364.2 | XP_005557421.1 | 4 | 1685 | 118 | |
| XM_005557372.2 | XP_005557429.1 | 3 | 1290 | 118 | ||
More Information
Set Membership: |
|

NR2F1
Genomic Map