Assay Details
Gene Symbol: ELANE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005587338.2 | XP_005587395.1 | 3 - 4 | 649 | 71 | |
XM_005587339.2 | XP_005587396.1 | 3 - 4 | 642 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
