Assay Details
| Gene Symbol: UCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564550.2 | XP_005564607.1 | 1 - 2 | 300 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005564550.2 transcripts.
Set Membership: |
|

UCN3
Genomic Map