Assay Details
Gene Symbol: ACOT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553905.2 | XP_005553962.1 | 1 - 2 | 334 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
