Assay Details
Gene Symbol: LOC102137053 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287711.1 | NP_001274640.1 | 4 - 5 | 349 | 82 | |
XM_005567098.2 | XP_005567155.1 | 4 - 5 | 566 | 82 | ||
GenBank mRNA | AB125163.1 | - | 4 - 5 | 407 | 82 | |
AB169480.1 | - | 4 - 5 | 349 | 82 | ||
AB179491.1 | - | 4 - 5 | 349 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
