Assay Details
Gene Symbol: ZNF706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005563823.2 | XP_005563880.1 | 2 - 3 | 759 | 70 | |
XM_005563824.2 | XP_005563881.1 | 2 - 3 | 155 | 70 | ||
XM_005563825.2 | XP_005563882.1 | 2 - 3 | 436 | 70 | ||
XM_005563826.2 | XP_005563883.1 | 2 - 3 | 153 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
