Assay Details
| Gene Symbol: CPT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283413.1 | NP_001270342.1 | 1 - 2 | 162 | 66 | |
| XM_015456928.1 | XP_015312414.1 | 1 - 2 | 141 | 66 | ||
| XM_015456929.1 | XP_015312415.1 | 2 - 3 | 634 | 66 | ||
| XM_015456930.1 | XP_015312416.1 | 2 - 3 | 523 | 66 | ||
| XM_015456931.1 | XP_015312417.1 | 2 - 3 | 634 | 66 | ||
| XM_015456932.1 | XP_015312418.1 | 2 - 3 | 634 | 66 | ||
| XM_015456933.1 | XP_015312419.1 | 2 - 3 | 240 | 66 | ||
| GenBank mRNA | AB168775.1 | - | 1 - 2 | 162 | 66 | |
More Information
Set Membership: |
|

CPT1B
Genomic Map