Assay Details
| Gene Symbol: DUSP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540903.2 | XP_005540960.1 | 3 - 4 | 2063 | 83 | |
| XM_005540904.2 | XP_005540961.1 | 3 - 4 | 2246 | 83 | ||
More Information
Set Membership: |
|

DUSP10
Genomic Map