Assay Details
| Gene Symbol: CENPW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551743.2 | XP_005551800.1 | 1 - 2 | 611 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CENPW| Gene Symbol: CENPW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551743.2 | XP_005551800.1 | 1 - 2 | 611 | 85 | |
Set Membership: |
|