Assay Details
| Gene Symbol: GCNT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559681.2 | XP_005559738.1 | 5 | 2346 | 77 | |
| XM_005559682.2 | XP_005559739.1 | 5 | 2116 | 77 | ||
| XM_005559683.2 | XP_005559740.1 | 4 | 2070 | 77 | ||
| XM_015452884.1 | XP_015308370.1 | 4 | 2439 | 77 | ||
More Information
Set Membership: |
|

GCNT3
Genomic Map