Assay Details
| Gene Symbol: B3GALT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005573375.2 | XP_005573432.1 | 5 | 1940 | 101 | |
| XM_005573379.2 | XP_005573436.1 | 4 | 1887 | 101 | ||
| XM_015432322.1 | XP_015287808.1 | 2 | 2314 | 101 | ||
More Information
Set Membership: |
|

B3GALT1
Genomic Map