Assay Details
Gene Symbol: SAPCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553484.2 | XP_005553541.1 | 1 - 2 | 373 | 99 | |
XM_015449615.1 | XP_015305101.1 | 1 - 2 | 373 | 99 | ||
XM_015449616.1 | XP_015305102.1 | 1 - 2 | 373 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
