Assay Details
Gene Symbol: TMEM91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589345.2 | XP_005589402.1 | 2 - 3 | 973 | 85 | |
XM_005589346.2 | XP_005589403.1 | 2 - 3 | 973 | 85 | ||
XM_005589347.2 | XP_005589404.1 | 2 - 3 | 973 | 85 | ||
XM_005589348.2 | XP_005589405.1 | 2 - 3 | 973 | 85 | ||
XM_005589349.2 | XP_005589406.1 | 2 - 3 | 973 | 85 | ||
XM_005589350.2 | XP_005589407.1 | 2 - 3 | 973 | 85 | ||
XM_005589351.2 | XP_005589408.1 | 2 - 3 | 973 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
