Assay Details
Gene Symbol: C3H7orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005548985.2 | XP_005549042.1 | 3 - 4 | 430 | 103 | |
GenBank mRNA | AB170407.1 | - | 4 - 5 | 590 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
