Assay Details
Gene Symbol: CYP11B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005564197.2 | XP_005564254.1 | 1 - 2 | 240 | 74 | |
XM_005564198.1 | XP_005564255.1 | 1 - 2 | 237 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
