Assay Details
| Gene Symbol: APOBEC3F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567253.2 | XP_005567310.1 | 7 | 1404 | 86 | |
| XM_015457184.1 | XP_015312670.1 | 7 | 1353 | 86 | ||
More Information
Set Membership: |
|

APOBEC3F
Genomic Map