Assay Details
Gene Symbol: C2H3orf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005548395.2 | XP_005548452.1 | 1 - 2 | 524 | 97 | |
XM_015446584.1 | XP_015302070.1 | 1 - 2 | 524 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
