Assay Details
| Gene Symbol: RPLP0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283935.1 | NP_001270864.1 | 7 - 8 | 860 | 69 | |
| GenBank mRNA | AB170149.1 | - | 7 - 8 | 860 | 69 | |
More Information
Set Membership: |
|

RPLP0
Genomic Map