Assay Details
Gene Symbol: CHST11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015431470.1 | XP_015286956.1 | 1 - 2 | 1365 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
