Assay Details
| Gene Symbol: KLHL25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560416.2 | XP_005560473.1 | 2 - 3 | 5233 | 85 | |
| XM_005560417.2 | XP_005560474.1 | 2 - 3 | 1969 | 85 | ||
| XM_005560418.2 | XP_005560475.1 | 2 - 3 | 2312 | 85 | ||
| XM_015453341.1 | XP_015308827.1 | 2 - 3 | 6025 | 85 | ||
| XM_015453342.1 | XP_015308828.1 | 2 - 3 | 5531 | 85 | ||
More Information
Set Membership: |
|

KLHL25
Genomic Map