Assay Details
Gene Symbol: AURKAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283503.1 | NP_001270432.1 | 2 - 3 | 918 | 53 | |
XM_015452204.1 | XP_015307690.1 | 3 - 4 | 808 | 53 | ||
XM_015452209.1 | XP_015307695.1 | 3 - 4 | 731 | 53 | ||
XM_015452212.1 | XP_015307698.1 | 3 - 4 | 860 | 53 | ||
GenBank mRNA | AB171069.1 | - | 2 - 3 | 918 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
