Assay Details
Gene Symbol: BOLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005542039.2 | XP_005542096.1 | 2 | 1291 | 106 | |
XM_005542040.2 | XP_005542097.1 | 2 | 1463 | 106 | ||
XM_005542041.2 | XP_005542098.1 | 2 | 1253 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
