Assay Details
| Gene Symbol: KCNIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287658.1 | NP_001274587.1 | 2 - 3 | 317 | 83 | |
| GenBank mRNA | AB169924.1 | - | 2 - 3 | 317 | 83 | |
More Information
Set Membership: |
|

KCNIP3
Genomic Map