Assay Details
| Gene Symbol: GNPNAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561269.2 | XP_005561326.1 | 3 - 4 | 473 | 110 | |
| XM_005561270.2 | XP_005561327.1 | 4 - 5 | 461 | 110 | ||
More Information
Set Membership: |
|

GNPNAT1
Genomic Map