Assay Details
| Gene Symbol: PTGES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015436347.1 | XP_015291833.1 | 2 - 3 | 326 | 86 | |
| GenBank mRNA | AY573809.1 | - | 2 - 3 | 211 | 86 | |
More Information
Set Membership: |
|

PTGES
Genomic Map