Assay Details
| Gene Symbol: PIGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540007.2 | XP_005540064.1 | 2 | 1249 | 84 | |
| XM_015445524.1 | XP_015301010.1 | 2 | 1056 | 84 | ||
More Information
Set Membership: |
|

PIGC
Genomic Map