Assay Details
Gene Symbol: CHST7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593385.2 | XP_005593442.1 | 1 - 2 | 1738 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
