Assay Details
Gene Symbol: EID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005559498.2 | XP_005559555.1 | 1 | 738 | 97 | |
GenBank mRNA | AB066547.1 | - | 905 | 97 | ||
AB170461.1 | - | 1 | 502 | 97 | ||
AB174396.1 | - | 1 | 541 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
