Assay Details
Gene Symbol: TSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005572892.2 | XP_005572949.1 | 1 - 2 | 805 | 114 | |
XM_005572893.2 | XP_005572950.1 | 1 - 2 | 805 | 114 | ||
XM_015431957.1 | XP_015287443.1 | 1 - 2 | 805 | 114 | ||
GenBank mRNA | AB170469.1 | - | 1 - 2 | 164 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
