Assay Details
| Gene Symbol: TSPAN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578122.2 | XP_005578179.1 | 6 - 7 | 764 | 64 | |
| XM_005578123.2 | XP_005578180.1 | 5 - 6 | 687 | 64 | ||
| XM_015434869.1 | XP_015290355.1 | 5 - 6 | 969 | 64 | ||
| XM_015434870.1 | XP_015290356.1 | 5 - 6 | 930 | 64 | ||
More Information
Set Membership: |
|

TSPAN18
Genomic Map