Assay Details
| Gene Symbol: TXNDC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543377.1 | XP_005543434.1 | 7 | 2017 | 151 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TXNDC12| Gene Symbol: TXNDC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543377.1 | XP_005543434.1 | 7 | 2017 | 151 | |
Set Membership: |
|