Assay Details
Gene Symbol: SMIM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544026.2 | XP_005544083.1 | 2 | 376 | 121 | |
XM_005544027.2 | XP_005544084.1 | 2 | 184 | 121 | ||
XM_005544028.2 | XP_005544085.1 | 2 | 689 | 121 | ||
XM_015438720.1 | XP_015294206.1 | 2 | 226 | 121 | ||
XM_015438728.1 | XP_015294214.1 | 2 | 1229 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
