Assay Details
| Gene Symbol: OPN1LW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283209.1 | NP_001270138.1 | 1 - 2 | 113 | 71 | |
| GenBank mRNA | AF158968.1 | - | 1 - 2 | 113 | 71 | |
| AF158975.1 | - | 1 - 2 | 113 | 71 | ||
More Information
Set Membership: |
|

OPN1LW
Genomic Map