Assay Details
| Gene Symbol: OPN1LW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283209.1 | NP_001270138.1 | 3 - 4 | 581 | 101 | |
| GenBank mRNA | AF158975.1 | - | 3 - 4 | 581 | 101 | |
More Information
Set Membership: |
|

OPN1LW
Genomic Map