Assay Details
| Gene Symbol: C2H3orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283716.1 | NP_001270645.1 | 3 - 4 | 263 | 59 | |
| GenBank mRNA | AB169301.1 | - | 3 - 4 | 263 | 59 | |
More Information
Set Membership: |
|

C2H3orf84
Genomic Map