Assay Details
| Gene Symbol: CCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283805.1 | NP_001270734.1 | 1 | 819 | 82 | |
| XM_015445357.1 | XP_015300843.1 | 6 | 82 | |||
| XM_015445358.1 | XP_015300844.1 | 5 | 82 | |||
| XM_015445359.1 | XP_015300845.1 | 5 | 82 | |||
| GenBank mRNA | AY065646.1 | - | 1 | 82 | ||
| AY065647.1 | - | 1 | 819 | 82 | ||
More Information
Set Membership: |
|

CCR3
Genomic Map