Assay Details
| Gene Symbol: METTL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283824.1 | NP_001270753.1 | 2 - 3 | 204 | 100 | |
| GenBank mRNA | AB174697.1 | - | 2 - 3 | 204 | 100 | |
More Information
Set Membership: |
|

METTL2A
Genomic Map