Assay Details
Gene Symbol: RAB8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285006.1 | NP_001271935.1 | 5 - 6 | 416 | 85 | |
XM_015440167.1 | XP_015295653.1 | 7 - 8 | 980 | 85 | ||
GenBank mRNA | AB169502.1 | - | 7 - 8 | 544 | 85 | |
AB172361.1 | - | 5 - 6 | 416 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
