Assay Details
| Gene Symbol: TAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285154.1 | NP_001272083.1 | 1 | 1117 | 78 | |
| GenBank mRNA | AB169639.1 | - | 1 | 1117 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAF7| Gene Symbol: TAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285154.1 | NP_001272083.1 | 1 | 1117 | 78 | |
| GenBank mRNA | AB169639.1 | - | 1 | 1117 | 78 | |
Set Membership: |
|