Assay Details
Gene Symbol: FCGR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283669.1 | NP_001270598.1 | 1 - 2 | 64 | 118 | |
XM_005541167.2 | XP_005541224.1 | 1 - 2 | 129 | 121 | ||
XM_005541168.2 | XP_005541225.1 | 1 - 2 | 130 | 118 | ||
XM_005541169.2 | XP_005541226.1 | 1 - 2 | 129 | 121 | ||
XM_015452315.1 | XP_015307801.1 | 1 - 2 | 129 | 121 | ||
XM_015452322.1 | XP_015307808.1 | 1 - 2 | 130 | 118 | ||
XM_015452330.1 | XP_015307816.1 | 1 - 2 | 129 | 121 | ||
GenBank mRNA | AF485813.1 | - | 1 - 2 | 64 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
