Assay Details
| Gene Symbol: SYT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285043.1 | NP_001271972.1 | 7 - 8 | 1108 | 99 | |
| GenBank mRNA | AB169684.1 | - | 7 - 8 | 1108 | 99 | |
More Information
Set Membership: |
|

SYT5
Genomic Map