Assay Details
| Gene Symbol: ZNF394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283169.1 | NP_001270098.1 | 2 - 3 | 786 | 93 | |
| XM_015446603.1 | XP_015302089.1 | 3 - 4 | 723 | 93 | ||
| XM_015446604.1 | XP_015302090.1 | 3 - 4 | 723 | 93 | ||
| GenBank mRNA | AB170885.1 | - | 2 - 3 | 789 | 93 | |
More Information
Set Membership: |
|

ZNF394
Genomic Map