Assay Details
Gene Symbol: CA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283685.1 | NP_001270614.1 | 4 - 5 | 524 | 65 | |
GenBank mRNA | AB174240.1 | - | 4 - 5 | 524 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
