Assay Details
Gene Symbol: SMIM43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555829.2 | XP_005555886.1 | 5 | 1155 | 78 | |
XM_005555830.2 | XP_005555887.1 | 5 | 1106 | 78 | ||
XM_005555831.2 | XP_005555888.1 | 5 | 1152 | 78 | ||
XM_005555834.2 | XP_005555891.1 | 5 | 1203 | 78 | ||
XM_005555835.2 | XP_005555892.1 | 4 | 1092 | 78 | ||
XM_015450789.1 | XP_015306275.1 | 3 | 861 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
