Assay Details
Gene Symbol: TSPAN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284120.1 | NP_001271049.1 | 5 - 6 | 970 | 60 | |
XM_015450499.1 | XP_015305985.1 | 5 - 6 | 1012 | 60 | ||
XM_015450500.1 | XP_015305986.1 | 4 - 5 | 859 | 60 | ||
GenBank mRNA | AB170093.1 | - | 5 - 6 | 968 | 60 | |
AB173982.1 | - | 5 - 6 | 970 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
