Assay Details
Gene Symbol: RHNO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284613.1 | NP_001271542.1 | 2 - 3 | 313 | 66 | |
GenBank mRNA | AB170663.1 | - | 2 - 3 | 313 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
