Assay Details
| Gene Symbol: SLC39A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284851.1 | NP_001271780.1 | 4 - 5 | 1059 | 97 | |
| GenBank mRNA | AB168166.1 | - | 4 - 5 | 1059 | 97 | |
More Information
Set Membership: |
|

SLC39A2
Genomic Map