Assay Details
| Gene Symbol: MRPL49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283495.1 | NP_001270424.1 | 3 - 4 | 383 | 72 | |
| GenBank mRNA | AB169536.1 | - | 3 - 4 | 397 | 72 | |
| AB170521.1 | - | 3 - 4 | 383 | 72 | ||
| AB174650.1 | - | 3 - 4 | 383 | 72 | ||
More Information
Set Membership: |
|

MRPL49
Genomic Map